Order Entry
Poland
ContactUsLinkComponent
VP 6-pole electrode socket, 20 m cable KA 19 (1 coaxial + 4 cords + 1 shielding) up to 130 C, free end 1 * 1 items
Catalog # SCOI285129810
Supplier:  SI Analytics
VP 6-pole electrode socket, 20 m cable KA 19 (1 coaxial + 4 cords + 1 shielding) up to 130 C, free end 1 * 1 items
Catalog # SCOI285129810
Supplier:  SI Analytics