DL-ALPHA-TOCOPHEROL METHOXYPOLYETHYLENE 1 * 50 mL
Catalog # SIAL733857-50ML
Supplier: Merck
Frequently Bought Together
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
n-Heptane, pure analytical reagent
Supplier: CHEMPUR
1-Chlorobutane ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
2-Ethoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone 99.5+% for analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals