Order Entry
Poland
Orders LinkContactUsLinkComponent
Caps and septa, BASIK™
Caps and septa, BASIK™
Snap cap and septa, non-slit, light blue, Soft-Guard™, White silicone/red PTFE, Light blue
Catalog # MICS9502C-E-10LB-M
Supplier:   MICROSOLV
Caps and septa, BASIK™
Catalog # MICS9502C-E-10LB-M
Supplier:  MICROSOLV
Caps and septa, BASIK™
Caps and septa, BASIK™
Snap cap and septa, non-slit, light blue, Soft-Guard™, White silicone/red PTFE, Light blue
Price
1 390 zł
Log in for availability
1 * 1.000 SZT
Product Details & Documents
For use with 12×32 mm autosampler vials.
Specifications
  • Septum:
    White silicone/red PTFE
  • Description:
    Snap cap and septa, non-slit, light blue, Soft-Guard™
  • Seq:
    10
  • Cap colour:
    Light blue