Caps and septa, BASIK™
Snap cap and septa, non-slit, black, Soft-Guard™, White silicone/red PTFE, Black
MICS9502C-E-10K
For use with 12×32 mm autosampler vials.
- Septum:White silicone/red PTFE
- Description:Snap cap and septa, non-slit, black, Soft-Guard™
- Seq:07
- Cap colour:Black