Caps and septa, BASIK™
Snap cap and septa, preassembled, Soft-Guard™, White silicone/red PTFE, Blue
numer katalogowy MICS9502C-E-10B
Dostawca: MICROSOLV
Przejdź do:
Product Details & Documents
For use with 12×32 mm autosampler vials.
Dane techniczne
- Septum:White silicone/red PTFE
- Description:Snap cap and septa, preassembled, Soft-Guard™
- Seq:01
- Cap colour:Blue