Order Entry
Poland
ContactUsLinkComponent
Sample holder, 1 inch round, 2/pk. Includes holder for 32 samples and holder for 24 samples. Suitable for use with Cary 7000 UMS or with UMA accessory on Cary 4000, 5000, and 6000i. For mounting 1 inch round solid samples for measurement by UV-Vis-NI 1 * 1 items
Catalog # HEWLG6876-60003
Supplier:  Agilent
Sample holder, 1 inch round, 2/pk. Includes holder for 32 samples and holder for 24 samples. Suitable for use with Cary 7000 UMS or with UMA accessory on Cary 4000, 5000, and 6000i. For mounting 1 inch round solid samples for measurement by UV-Vis-NI 1 * 1 items
Catalog # HEWLG6876-60003
Supplier:  Agilent
Frequently Bought Together