ICH/USP oral target elements standard B, Ni 200µg/mL, Ag and Se 150 µg/mL, V 100 µg/mL, Tl 8 µg/mL, Co 5 µg/mL in 2 % HNO4 1 * 1 items
Catalog # HEWL5190-9767
Supplier: Agilent
ICH/USP oral target elements standard B, Ni 200µg/mL, Ag and Se 150 µg/mL, V 100 µg/mL, Tl 8 µg/mL, Co 5 µg/mL in 2 % HNO4 1 * 1 items
Catalog # HEWL5190-9767
Supplier: Agilent
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection