TANTALUM 1 µG/ML H20/TR PURE 125 ML 1 * 125 mL
Catalog # PERCN9304271
Supplier: PerkinElmer
Frequently Bought Together
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Powder Pillows, water hardness, ManVer 2
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach
Iron standard solution, for ICP-MS, Pure Plus grade
Supplier: PerkinElmer