Order Entry
Poland
Orders LinkContactUsLinkComponent
Mag-Bind® cfDNA kit

Mag-Bind® cfDNA kit

Mag-Bind® cfDNA Kit, 200
Catalog # M3298-02
Supplier:   OMEGA BIO-TEK
Mag-Bind® cfDNA kit
Catalog # M3298-02
Supplier:  OMEGA BIO-TEK
CAS Number:  
Mag-Bind® cfDNA kit
Mag-Bind® cfDNA kit

Mag-Bind® cfDNA Kit, 200

CAS Number:  
Price
12 240 zł
Log in for availability
1 * 200 SZT
Product Details & Documents

The Mag-Bind® cfDNA Kit is designed for rapid and reliable isolation of circulating DNA from 500 to 4000 µl plasma or serum samples. The Mag-Bind® cfDNA Kit can be processed manually or with automated platforms. The procedure eliminates the need for funnels and vacuum steps, providing hands-free operation in automated protocols.

  • Optimum recovery of cell-free DNA with minimal genomic DNA contamination
  • Versatile sample input volumes (500 to 4000 µl of serum/plasma) and low elution volumes (50 µl)
  • High quality DNA suitable for a variety of downstream applications such as qPCR and NGS
  • Reduced binding and lysis buffers allow for larger sample volume processing

The uniquely formulated binding buffer allows for large sample volumes to be processed in automated formats with 4 ml of serum and plasma being processed in 24-well plates. The magnetic response of the Mag-Bind® Particles CH allow for fast magnetisation during steps requiring high volumes and the high binding capacity allows for reduced amounts of magnetic particles to be required, thus reducing the elution volume required. 4 ml serum or plasma can be eluted in 50 µl.

The system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with a unique binding system that targets smaller DNA fragments (150 to 400 bp) and minimises the binding or larger fragments such as gDNA.

Specifications

Specifications

  • Pk:
    200
  • Description:
    Mag-Bind® cfDNA Kit
  • Cat. No.:
    M3298-02
  • Prep size:
    Scalable
  • Refine target molecule:
    cfDNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Description:
    Mag-Bind® cfDNA Kit
  • Sample type:
    Biofluids (serum, plasma, saliva)
  • Sample size:
    0,5 - 4 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.