Order Entry
Poland
ContactUsLinkComponent
Cynomolgus Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Ac cession XP005544945.1). In the region Leu 24 - Gln 186, the AA sequence of Cynomolgus and Rhesus macaque 4-1BB are ho 1 * 1 mg
Catalog # ACRB41B-C52H4-1MG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Ac cession XP005544945.1). In the region Leu 24 - Gln 186, the AA sequence of Cynomolgus and Rhesus macaque 4-1BB are ho 1 * 1 mg
Catalog # ACRB41B-C52H4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together