Order Entry
Poland
ContactUsLinkComponent
CONVERTIBLE SAPPHIRE INJECTOR. HF-RESISTANT. THIS SAPPHIRE INJECTOR IS USED FOR ENVIRONMENTAL, GEOCHEMISTRY, AND HF APPLICATIONS. TH IS INJECTOR INCLUDES THE BLACK PEEK ADAPTER (N0782020) TO CONVERT FOR USE WITH OPTIMA 4000/5000/7000 SERIES INSTRUMEN 1 * 1 items
Catalog # PERCN0782006
Supplier:  PerkinElmer
CONVERTIBLE SAPPHIRE INJECTOR. HF-RESISTANT. THIS SAPPHIRE INJECTOR IS USED FOR ENVIRONMENTAL, GEOCHEMISTRY, AND HF APPLICATIONS. TH IS INJECTOR INCLUDES THE BLACK PEEK ADAPTER (N0782020) TO CONVERT FOR USE WITH OPTIMA 4000/5000/7000 SERIES INSTRUMEN 1 * 1 items
Catalog # PERCN0782006
Supplier:  PerkinElmer
Frequently Bought Together