Order Entry
Poland
ContactUsLinkComponent
CONSUMABLE SCREEN FOR USE ON THE DIRECT SAMPLE ANALYSIS (DSA) ION SOURCE FOR AXION 2 TOF MS. FOR HOLDING LIQUID SAMPLES. USE WITH DS A MESH HOLDER ASSEMBLY (MZ300579). 1 * 1 items
Catalog # PERCMZ305382
Supplier:  PerkinElmer
CONSUMABLE SCREEN FOR USE ON THE DIRECT SAMPLE ANALYSIS (DSA) ION SOURCE FOR AXION 2 TOF MS. FOR HOLDING LIQUID SAMPLES. USE WITH DS A MESH HOLDER ASSEMBLY (MZ300579). 1 * 1 items
Catalog # PERCMZ305382
Supplier:  PerkinElmer
Frequently Bought Together