Order Entry
Poland
ContactUsLinkComponent
Drop-n-Stain™ CF640R donkey anti-rabbit IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting.  To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamste 1 * 5 mL
Catalog # BTIU20962
Supplier:  Biotium
Drop-n-Stain™ CF640R donkey anti-rabbit IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting.  To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamste 1 * 5 mL
Catalog # BTIU20962
Supplier:  Biotium
Frequently Bought Together