30 components 10mg/l each of Ag Al As Ba Be Bi Ca Cd Co Cr Cs Cu Fe Ga Hg In K Li Mg Mn Na Ni Pb Rb Se Sr Tl V U Zn in HNO3 5% . Duree de conservation : 6 mois a ouverture du flacon, Duree de 1 * 100 mL
Catalog # ACSD8125.10.5NL1.1
Supplier: CPACHEM
Frequently Bought Together