Order Entry
Poland
ContactUsLinkComponent
Drop-n-Stain CF488A goat anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting. T o minimize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 5 mL
  BTIU20956
 :  Biotium
Drop-n-Stain CF488A goat anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting. T o minimize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 5 mL
  BTIU20956
 :  Biotium
Frequently Bought Together