CONOSTAN S-21 100 ppm +Co+K+Li+W 75cSt,2X200G 1 * 200 g
Catalog # PROOCON151-021-007
Supplier: LGC Standards PROMOCHEM
CONOSTAN S-21 100 ppm +Co+K+Li+W 75cSt,2X200G 1 * 200 g
Catalog # PROOCON151-021-007
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
Methanol UHPLC for mass spectrometry 1 * 1 L
Supplier: Merck
n-Heptane, pure
Supplier: CHEMPUR
Tripalmitin (glycerol tripalmitate), Sigma-Aldrich®
Supplier: Merck
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl nitrite, Sigma-Aldrich®
Supplier: Merck
1-Butanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
(±)-1,2-Propanediol ACS
Supplier: Thermo Fisher Scientific