Order Entry
Poland
ContactUsLinkComponent
Includes 3 stainless steel mounting hooks and 3 carabiners. Hooks mount to rear grille and carabiners secure power cords and tubing along rear wall to keep them contained and away from work area. For use with Purifier Class II Biosafety Cabinets.EA 1 * 1 items
Catalog # LBCP3859100
Supplier:  LABCONCO
Includes 3 stainless steel mounting hooks and 3 carabiners. Hooks mount to rear grille and carabiners secure power cords and tubing along rear wall to keep them contained and away from work area. For use with Purifier Class II Biosafety Cabinets.EA 1 * 1 items
Catalog # LBCP3859100
Supplier:  LABCONCO
Frequently Bought Together