Order Entry
Poland
ContactUsLinkComponent
Aniline, 10X in acetate buffer is designed to use as catalytic reagent for the optimal coupling of protein carbonyls with aminooxy compounds. 1 * 10 mL
Catalog # 91057.
Supplier:  Biotium
Aniline, 10X in acetate buffer is designed to use as catalytic reagent for the optimal coupling of protein carbonyls with aminooxy compounds. 1 * 10 mL
Catalog # 91057.
Supplier:  Biotium
Frequently Bought Together