Order Entry
Poland
ContactUsLinkComponent
Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondria l ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochon 1 * 100 µG
Catalog # BWRLBS61212
Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondria l ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochon 1 * 100 µG
Catalog # BWRLBS61212
Frequently Bought Together