Order Entry
Poland
ContactUsLinkComponent
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-mouse IgG (H+L) with CF™535ST for (d)-STORM supe r-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, r 1 * 50 µl
Catalog # 20821-50UL
Supplier:  Biotium
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-mouse IgG (H+L) with CF™535ST for (d)-STORM supe r-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, r 1 * 50 µl
Catalog # 20821-50UL
Supplier:  Biotium
Frequently Bought Together