Order Entry
Poland
ContactUsLinkComponent
High purity Adenosine 5’-triphosphate for use as a cofactor for research, biochemical enzyme assays and in vitro diagnostic analysis. Adenosine 5’-triphosphate, disodium salt (ATP, 5’-ATP, ATP-Na2). MW: 507.18 for free acid. Purity: 97.9% ATP conten 1 * 100 g
  MEGAC-ATP-100G
 :  MEGAZYME
High purity Adenosine 5’-triphosphate for use as a cofactor for research, biochemical enzyme assays and in vitro diagnostic analysis. Adenosine 5’-triphosphate, disodium salt (ATP, 5’-ATP, ATP-Na2). MW: 507.18 for free acid. Purity: 97.9% ATP conten 1 * 100 g
  MEGAC-ATP-100G
 :  MEGAZYME
Frequently Bought Together