Order Entry
Poland
Orders LinkContactUsLinkComponent
4-(2-(5-Bromo-2-oxoindolin-3-ylidene)hydrazinyl)benzenesulphonamide ≥97% Cdk2 inhibitor

4-(2-(5-Bromo-2-oxoindolin-3-ylidene)hydrazinyl)benzenesulphonamide ≥97% Cdk2 inhibitor

Catalog # ABCAAB145053-5
Supplier:   Abcam
4-(2-(5-Bromo-2-oxoindolin-3-ylidene)hydrazinyl)benzenesulphonamide ≥97% Cdk2 inhibitor
Catalog # ABCAAB145053-5
Supplier:  Abcam
CAS Number:  222035-13-4
4-(2-(5-Bromo-2-oxoindolin-3-ylidene)hydrazinyl)benzenesulphonamide ≥97% Cdk2 inhibitor
CAS Number:  222035-13-4
Price
1 680 zł
Log in for availability
1 * 5 mg
Product Details & Documents

Potent, selective, ATP-competitive Cdk2 inhibitor (IC₅₀ = 60 nM). Cell-permeable.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  CDK2-IN-3
  • CAS Number:222035-13-4
  • Molecular Weight:395,23 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% SC 221409, CDK2 inhibitor II