Order Entry
Poland
ContactUsLinkComponent
White Crystalline Powder.Assay: 98.5-101.5% Specific rotation: +6.3 to + 7.3. Loss on drying: 0.3%.Residue on ignition: 0.3% ,Chloride: 0.5%,Sulfate: 0.03%,Iron: 0.003%,Heavy metals: 0.0015%Organic volatile 1 * 500 g
Catalog # ICNA0219960890
Supplier:  MP Biomedicals
White Crystalline Powder.Assay: 98.5-101.5% Specific rotation: +6.3 to + 7.3. Loss on drying: 0.3%.Residue on ignition: 0.3% ,Chloride: 0.5%,Sulfate: 0.03%,Iron: 0.003%,Heavy metals: 0.0015%Organic volatile 1 * 500 g
Catalog # ICNA0219960890
Supplier:  MP Biomedicals
Frequently Bought Together