Order Entry
Poland
ContactUsLinkComponent
AGAROSE. Molecular Biology ReagentIdeal for nucleic acid electrophoresis, analysis and purification.EEO: 0.09-0.13Moisture: No detec table DNase, RNase, or protease. 1 * 10 g
Catalog # ICNA0219398310
Supplier:  MP Biomedicals
AGAROSE. Molecular Biology ReagentIdeal for nucleic acid electrophoresis, analysis and purification.EEO: 0.09-0.13Moisture: No detec table DNase, RNase, or protease. 1 * 10 g
Catalog # ICNA0219398310
Supplier:  MP Biomedicals
Frequently Bought Together