0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
pH combination electrode, gel-filled, pH 0…14, 0 … 80°C 1x ceramic diaphragm, Polycarbonate/Glass body, 1 m cable, BNC connector 1 * 1 items
HACHLZW5050970002
:
Hach
pH combination electrode, gel-filled, pH 0…14, 0 … 80°C 1x ceramic diaphragm, Polycarbonate/Glass body, 1 m cable, BNC connector 1 * 1 items
HACHLZW5050970002
:
Hach
Frequently Bought Together
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
: VWR Collection
pH electrodes, SenTix®
: WTW
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Bench Protectors
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Nitrile gloves, TouchNTuff® 92-600 / 92-605
: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
: KIMBERLY CLARK
Combined pH electrodes, sensION+
: Hach
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF
VWR®, Latex Gloves
: VWR Collection