SILICA GEL ORANGE (COBALT FREE) 1 * 5 kg
Catalog # CHMP159055810.5000
Dostawca: CHEMPUR
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Dostawca: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dostawca: Merck
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Dostawca: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Dostawca: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dostawca: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Dostawca: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Dostawca: VWR Chemicals
Isoamyl alcohol, pure analytical reagent
Dostawca: CHEMPUR
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals