Order Entry
Poland
ContactUsLinkComponent
A potent, selective, brain permeable inhibitor of GSK3 and GSK3 (Kis = 6.9 and 31 nM, respectively) 1 * 1 mg
Catalog # CAYM16676-1
Supplier:  Cayman Chemical
A potent, selective, brain permeable inhibitor of GSK3 and GSK3 (Kis = 6.9 and 31 nM, respectively) 1 * 1 mg
Catalog # CAYM16676-1
Supplier:  Cayman Chemical
Frequently Bought Together