Order Entry
Poland
ContactUsLinkComponent
A non-competitive, cell permeable inhibitor of calcium transport by SERCAs (IC50 values are cell type-dependent and range from 2-80 nM) 1 * 10 mg
Catalog # CAYM10522-10
Supplier:  Cayman Chemical
A non-competitive, cell permeable inhibitor of calcium transport by SERCAs (IC50 values are cell type-dependent and range from 2-80 nM) 1 * 10 mg
Catalog # CAYM10522-10
Supplier:  Cayman Chemical
Frequently Bought Together