Order Entry
Poland
ContactUsLinkComponent
A fluoroquinolone antibiotic that displays broad spectrum antibacterial activity by inhibiting the bacterial regulatory enzyme DNA g yrase (IC50 = 0.92 µg/ml) as well as topoisomerase IV (IC50 = 1.62 µg/ml) 1 * 1 g
Catalog # CAYM16923-1
Supplier:  Cayman Chemical
A fluoroquinolone antibiotic that displays broad spectrum antibacterial activity by inhibiting the bacterial regulatory enzyme DNA g yrase (IC50 = 0.92 µg/ml) as well as topoisomerase IV (IC50 = 1.62 µg/ml) 1 * 1 g
Catalog # CAYM16923-1
Supplier:  Cayman Chemical
Frequently Bought Together