Order Entry
Poland
Orders LinkContactUsLinkComponent
2-Ethoxyethyl acetate

2-Ethoxyethyl acetate

4 L
Catalog # SPCME1024-4LTGL
Flammable
Health
Irritant
Supplier:   Spectrum Chemical
2-Ethoxyethyl acetate
Catalog # SPCME1024-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  111-15-9
2-Ethoxyethyl acetate

4 L

CAS Number:  111-15-9
Price
1 550 zł
Log in for availability
1 * 4 L
Product Details & Documents
Synonyms:  Ethylglycol acetate
  • CAS Number:111-15-9
  • Formula:C₆H₁₂O₃
  • MDL Number:MFCD00009251
  • Molecular Weight:132.16 g/mol
  • Merck Index:13,03787
  • Boiling Pt:156 °C (1013 hPa)
  • Melting Pt:–62 °C
  • Density:0.976 g/cm³ (20 °C)
  • Flash Pt:54 °C
  • EINECS:203-839-2
  • UN:1172
  • ADR:3,III
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pk:
    4 L

Specifications