Octan 2-etoksyetylu
200 L
Catalog # SPCME1024-200LTBL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Ester etoksyetylowy kwasu octowego, Octan etyloglikolu
- CAS Number:111-15-9
- Formula:C₆H₁₂O₃
- MDL Number:MFCD00009251
- Molecular Weight:132.16 g/mol
- Merck Index:13,03787
- Boiling Pt:156 °C (1013 hPa)
- Melting Pt:–62 °C
- Density:0,976 g/cm³ (20 °C)
- Flash Pt:54 °C
- EINECS:203-839-2
- UN:1172
- ADR:3,III
Specifications
Specification Test Results
- Assay:99% Min.
Specifications
- Ilość:200 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



