Order Entry
Poland
Orders LinkContactUsLinkComponent
Ethanolamine NF

Ethanolamine NF

200 L
Catalog # SPCMM1322-200LTBL
Corrosive
Irritant
Supplier:   Spectrum Chemical
Ethanolamine NF
Catalog # SPCMM1322-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  141-43-5
Ethanolamine NF
Ethanolamine NF

200 L

CAS Number:  141-43-5
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:13,03762
  • Boiling Pt:171 °C (1013 hPa)
  • Melting Pt:10.5 °C
  • Density:1.0164 g/cm³ (20 °C)
  • Flash Pt:93 °C
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pk:
    200 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.