Order Entry
Poland
Orders LinkContactUsLinkComponent
1,3-Dihydroxyacetone dimer ≥98%

1,3-Dihydroxyacetone dimer ≥98%

Fiber drum, 25 kg
Catalog # SPCMD1038-25KGBL
Irritant
Supplier:   Spectrum Chemical
1,3-Dihydroxyacetone dimer ≥98%
Catalog # SPCMD1038-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  62147-49-3
1,3-Dihydroxyacetone dimer ≥98%

Fiber drum, 25 kg

CAS Number:  62147-49-3
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents

Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
  • CAS Number:62147-49-3
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00051019
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Assay:
    98% min.

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.