1,3-Dihydroxyacetone dimer ≥98%
Glass, amber, 1 kg
Catalog # SPCMD1038-1KG
Supplier: Spectrum Chemical
1,3-Dihydroxyacetone dimer ≥98%
Catalog # SPCMD1038-1KG
Supplier: Spectrum Chemical
CAS Number: 62147-49-3
Jump to:
Product Details & Documents
Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonyms: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
- CAS Number:62147-49-3
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00051019
- Molecular Weight:180.16 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Assay:98% min.
Specifications
- Pack type:Glass, amber
- Pk:1 kg



