Order Entry
Poland
ContactUsLinkComponent
In addition to angle of rotation and Brix, the RePo-4 also converts and displays the amount of fructose (%) and honey moisture (%). The RePo-4 is ideal for total quality control of honey. 1 * 1 items
Catalog # 635-0749
Supplier:  Atago
undefined
In addition to angle of rotation and Brix, the RePo-4 also converts and displays the amount of fructose (%) and honey moisture (%). The RePo-4 is ideal for total quality control of honey. 1 * 1 items
Catalog # 635-0749
Supplier:  Atago
Frequently Bought Together