Order Entry
Poland
Orders LinkContactUsLinkComponent
Zanamivir, Reference Standards
Zanamivir, Reference Standards
Zanamivir Related Compounds Mixture, USP reference standard, Vial, 2,5 mg
Catalog # 1724077.
Supplier:   USP
Zanamivir, Reference Standards
Catalog # 1724077.
Supplier:  USP
CAS Number:  
Zanamivir, Reference Standards
Zanamivir Related Compounds Mixture, USP reference standard, Vial, 2,5 mg
CAS Number:  
Price
6 250 zł
Log in for availability
1 * 2,5 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zanamivir Resolution Mixture: Mixture of zanamivir and talo-zanamivir
 : US Pharmacopea Reference Standard
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    2,5 mg
  • Standards:
    USP reference standard
  • Description:
    Zanamivir Related Compounds Mixture
Specifications