LIDOCAINE N-OXIDE (2-(DIETHYLOXIDOAMINO)-N-(2,6-DIMETHYLPHENYL)ACETAMIDE) 1 * 10 mg
PROOLGCFOR0102.01
LIDOCAINE N-OXIDE (2-(DIETHYLOXIDOAMINO)-N-(2,6-DIMETHYLPHENYL)ACETAMIDE) 1 * 10 mg
PROOLGCFOR0102.01
Frequently Bought Together
pH buffers, ready to use
: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
Conductivity standards
: HAMILTON BONADUZ
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR®, Flow Cup viscosity standards
: VWR Chemicals