SULFUR - S @ 3.00WT% IN LIGHT MINERAL OIL, 500ML 1 * 500 mL
PROOVHGS20MIN3P500
SULFUR - S @ 3.00WT% IN LIGHT MINERAL OIL, 500ML 1 * 500 mL
PROOVHGS20MIN3P500
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection