TEMAZEPAM-D5 0.1 MG/ML IN METHANOL 1 * 1 mL
Catalog # PROOAMP0087.80-02
Supplier: LGC Standards PROMOCHEM
TEMAZEPAM-D5 0.1 MG/ML IN METHANOL 1 * 1 mL
Catalog # PROOAMP0087.80-02
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ