Order Entry
Poland
ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, A, WB, 230V. . Elec. lift, B-305 bath, SJ 29/32 Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300251A011
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ29/32, A, WB, 230V. . Elec. lift, B-305 bath, SJ 29/32 Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300251A011
Supplier:  BUCHI
Frequently Bought Together