Order Entry
Poland
ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, V, P+G, WB, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Vertical (V) P+G coating, int.: Wou lff b., w/o int. 1 * 1 items
Catalog # BUCH11R300211V111
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ29/32, V, P+G, WB, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Vertical (V) P+G coating, int.: Wou lff b., w/o int. 1 * 1 items
Catalog # BUCH11R300211V111
Supplier:  BUCHI
Frequently Bought Together