Rotavapor R-300 no bath, SJ24/40, A, P+G, I-300P, 230V. . Lift System: Elec. Heating Bath: w/o SJ 24/40 Glass set: Diagonal (A) P+G coating, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300202A131
Supplier: BUCHI
Frequently Bought Together