Order Entry
Poland
ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, C, I-300P, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Cold trap, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300151C031
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ29/32, C, I-300P, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Cold trap, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300151C031
Supplier:  BUCHI
Frequently Bought Together