Rotavapor R-300 B-301, SJ29/32, A, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass s et: Diagonal (A) P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
numer katalogowy BUCH11R300111A151
Supplier: BUCHI
Frequently Bought Together