Propargyl chloride 98%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Propargyl chloride 98%
Catalog # ACRO451840050
Supplier: Thermo Fisher Scientific
CAS Number: 624-65-7
Jump to:
Product Details & Documents
Synonyms: 3-Chloropropyne, 3-Chloro-1-propyne
- CAS Number:624-65-7
- Formula:C₃H₃Cl
- MDL Number:MFCD00000980
- Molecular Weight:74.51 g/mol
- Boiling Pt:58…61 °C (760 mmHg)
- Melting Pt:–77 °C
- Density:1.03 g/cm³
- Flash Pt:19 °C
- Storage Temperature:Refrigerator
- UN:3286
- ADR:3,II
Documents
ACRO45184
SDS
Specifications
Specification Test Results
- Appearance:Colorless to yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4370 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



