Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonium iodide 99%, extra pure

Ammonium iodide 99%, extra pure

Glass bottle, 100 g
Catalog # ACRO448071000
Irritant
Ammonium iodide 99%, extra pure
Catalog # ACRO448071000
CAS Number:  12027-06-4
Ammonium iodide 99%, extra pure
Ammonium iodide 99%, extra pure

Glass bottle, 100 g

CAS Number:  12027-06-4
Price
381 zł
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:12027-06-4
  • Formula:NH₄I
  • MDL Number:MFCD00011424
  • Molecular Weight:144.94 g/mol
  • Melting Pt:551 °C (subl.)
  • Density:2.51 g/cm³
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder (May turn yellow on storage)
  • Titration Argentometric:
    98.5 to 101.5 %
  • Residue after ignition:
    ≤0.05 %
  • Heavy metals (as Pb):
    ≤0.001 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.