Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonium iodide 99%, extra pure

Ammonium iodide 99%, extra pure

Glass bottle, 2,5 kg
Catalog # ACRO448070025
Irritant
Ammonium iodide 99%, extra pure
Catalog # ACRO448070025
CAS Number:  12027-06-4
Ammonium iodide 99%, extra pure
Ammonium iodide 99%, extra pure

Glass bottle, 2,5 kg

CAS Number:  12027-06-4
Price
4 500 zł
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:12027-06-4
  • Formula:NH₄I
  • MDL Number:MFCD00011424
  • Molecular Weight:144.94 g/mol
  • Melting Pt:551 °C (subl.)
  • Density:2.51 g/cm³
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder (May turn yellow on storage)
  • Titration Argentometric:
    98.5 to 101.5 %
  • Residue after ignition:
    ≤0.05 %
  • Heavy metals (as Pb):
    ≤0.001 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 kg

Specifications