Order Entry
Poland
Orders LinkContactUsLinkComponent
Aluminium acetate basic 90% | Avantor

Aluminium acetate basic 90%

Glass bottle, 1 kg
Catalog # ACRO428190010
Irritant
Aluminium acetate basic 90%
Catalog # ACRO428190010
CAS Number:  142-03-0
Aluminium acetate basic 90%
Aluminium acetate basic 90%

Glass bottle, 1 kg

CAS Number:  142-03-0
Price
907 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Aluminium Acetate Monobasic
  • CAS Number:142-03-0
  • Formula:C₄H₇AlO₅
  • MDL Number:MFCD00008688
  • Storage Temperature:Temperatura otoczenia

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Aluminium (Al):
    15.0 to 18.3 % (ICP)

Specifications

  • Rodzaj opakowania:
    Glass bottle
  • Ilość:
    1 kg

Specifications