Linoleic acid 60% tech.
Plastic drum, 2,5 kg
Supplier: Thermo Fisher Scientific
Linoleic acid 60% tech.
Catalog # ACRO413190025
Supplier: Thermo Fisher Scientific
CAS Number: 60-33-3
Jump to:
Product Details & Documents
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- CAS Number:60-33-3
- Formula:C₁₈H₃₂O₂
- MDL Number:MFCD00064241
- Molecular Weight:280.45 g/mol
- Boiling Pt:229…230 °C (16 mmHg)
- Melting Pt:–5 °C
- Density:0.9 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Refrigerator
- EINECS:200-470-9
Documents
ACRO41319
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≤3 % C18 (sat.)
- Acid value:195 to 205 mg KOH/g
- Iodine value:130 to 148 g I2/100 g
Specifications
- Pack type:Plastic drum
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



