1,2-Benzenedithiol 96%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
1,2-Benzenedithiol 96%
Catalog # ACRO334920050
Supplier: Thermo Fisher Scientific
CAS Number: 17534-15-5
Jump to:
Product Details & Documents
Synonyms: Benzene-1,2-dithiol
- CAS Number:17534-15-5
- Formula:C₆H₆S₂
- MDL Number:MFCD00004835
- Molecular Weight:142.25 g/mol
- Boiling Pt:52 - 54 °C (0,1 -rr)
- Melting Pt:27…28 °C
- Density:1,236 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:241-530-4
Documents
ACRO33492
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid or solid
- Infrared spectrum:Conforms
- GC:≥95 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



